Anti-RBP5 Mouse Monoclonal Antibody [clone: 2D1]

Lieferant: Abnova
ABNOH00083758-M05EA 382 EUR
ABNOH00083758-M05
Anti-RBP5 Mouse Monoclonal Antibody [clone: 2D1]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant RBP5.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RBP5
Clonality: Monoclonal
Clone: 2D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR