Anti-CRELD1 Rabbit Polyclonal Antibody
Lieferant: Abnova
ABNOH00078987-D01EA 380 EUR
ABNOH00078987-D01
Anti-CRELD1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human CRELD1 protein.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CRELD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human