Anti-CLEC2D Mouse Monoclonal Antibody [clone: 2E11]

Lieferant: Abnova
ABNOH00029121-M03EA 382 EUR
ABNOH00029121-M03
Anti-CLEC2D Mouse Monoclonal Antibody [clone: 2E11]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant CLEC2D.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CLEC2D
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR