Anti-IGKV1OR2-108 Mouse Monoclonal Antibody [clone: 3A11]

Lieferant: Abnova
ABNOH00028862-M01EA 394 EUR
ABNOH00028862-M01
Anti-IGKV1OR2-108 Mouse Monoclonal Antibody [clone: 3A11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant IGKV1OR2-108.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: IGKV1OR2-108
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR