Anti-PCDH11X Mouse Monoclonal Antibody [clone: 7B4]

Lieferant: Abnova
ABNOH00027328-M05EA 394 EUR
ABNOH00027328-M05
Anti-PCDH11X Mouse Monoclonal Antibody [clone: 7B4]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PCDH11X.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PCDH11X
Clonality: Monoclonal
Clone: 7B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR