Anti-FIGN Mouse Monoclonal Antibody [clone: 2F8]

Lieferant: Abnova
ABNOH00055137-M02EA 382 EUR
ABNOH00055137-M02
Anti-FIGN Mouse Monoclonal Antibody [clone: 2F8]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant FIGN.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FIGN
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR