Anti-HOOK1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00051361-D01EA 368 EUR
ABNOH00051361-D01
Anti-HOOK1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human HOOK1 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HOOK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR