Anti-TBC1D1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00023216-D01EA 368 EUR
ABNOH00023216-D01
Anti-TBC1D1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human TBC1D1 protein.

  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TBC1D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR