Anti-FGF21 Mouse Monoclonal Antibody [clone: 3E20]

Lieferant: Abnova
ABNOH00026291-M06AEA 382 EUR
ABNOH00026291-M06A
Anti-FGF21 Mouse Monoclonal Antibody [clone: 3E20]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant FGF21.

Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 3E20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR