Anti-GIPC2 Mouse Monoclonal Antibody [clone: 4H8]

Lieferant: Abnova
ABNOH00054810-M02EA 382 EUR
ABNOH00054810-M02
Anti-GIPC2 Mouse Monoclonal Antibody [clone: 4H8]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant GIPC2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GIPC2
Clonality: Monoclonal
Clone: 4H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR