Anti-ZBTB1 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00022890-B01EA 394 EUR
ABNOH00022890-B01
Anti-ZBTB1 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human ZBTB1 protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ZBTB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR