Anti-PLA2G3 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00050487-D01EA 380 EUR
ABNOH00050487-D01
Anti-PLA2G3 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human PLA2G3 protein.

  • Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PLA2G3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR