Anti-GNA14 Mouse Monoclonal Antibody [clone: 2H8]

Lieferant: Abnova
ABNOH00009630-M06AEA 382 EUR
ABNOH00009630-M06A
Anti-GNA14 Mouse Monoclonal Antibody [clone: 2H8]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant GNA14.

Type: Primary
Antigen: GNA14
Clonality: Monoclonal
Clone: 2H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR