Anti-RAB36 Mouse Monoclonal Antibody [clone: 6A6]

Lieferant: Abnova
ABNOH00009609-M01AEA 382 EUR
ABNOH00009609-M01A
Anti-RAB36 Mouse Monoclonal Antibody [clone: 6A6]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant RAB36.

Type: Primary
Antigen: RAB36
Clonality: Monoclonal
Clone: 6A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR