Anti-STIP1 Mouse Monoclonal Antibody [clone: 1C6]
Lieferant: Abnova
ABNOH00010963-M06EA 394 EUR
ABNOH00010963-M06
Anti-STIP1 Mouse Monoclonal Antibody [clone: 1C6]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant STIP1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: STIP1
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat