Anti-APOBEC2 Mouse Monoclonal Antibody [clone: 4D7]

Lieferant: Abnova
ABNOH00010930-M01EA 382 EUR
ABNOH00010930-M01
Anti-APOBEC2 Mouse Monoclonal Antibody [clone: 4D7]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant APOBEC2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: APOBEC2
Clonality: Monoclonal
Clone: 4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR