Anti-PTTG1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00009232-D02PEA 368 EUR
ABNOH00009232-D02P
Anti-PTTG1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human PTTG1 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: PTTG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR