Anti-PLA2G10 Mouse Monoclonal Antibody [clone: 5G11]

Lieferant: Abnova
ABNOH00008399-M01EA 382 EUR
ABNOH00008399-M01
Anti-PLA2G10 Mouse Monoclonal Antibody [clone: 5G11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PLA2G10.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PLA2G10
Clonality: Monoclonal
Clone: 5G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR