Anti-USP10 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00009100-B01PEA 394 EUR
ABNOH00009100-B01P
Anti-USP10 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human USP10 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: USP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human