Anti-SYT7 Mouse Monoclonal Antibody [clone: 4H4]

Lieferant: Abnova
ABNOH00009066-M01EA 382 EUR
ABNOH00009066-M01
Anti-SYT7 Mouse Monoclonal Antibody [clone: 4H4]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant SYT7.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SYT7
Clonality: Monoclonal
Clone: 4H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR