Anti-SLC19A2 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00010560-B01PEA 394 EUR
ABNOH00010560-B01P
Anti-SLC19A2 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human SLC19A2 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: SLC19A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human