Anti-TFEB Mouse Monoclonal Antibody [clone: S2]

Lieferant: Abnova
ABNOH00007942-M02EA 394 EUR
ABNOH00007942-M02
Anti-TFEB Mouse Monoclonal Antibody [clone: S2]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant TFEB.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TFEB
Clonality: Monoclonal
Clone: S2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR