Anti-PRR4 Mouse Monoclonal Antibody [clone: 1D2]

Lieferant: Abnova
ABNOH00011272-M02EA 382 EUR
ABNOH00011272-M02
Anti-PRR4 Mouse Monoclonal Antibody [clone: 1D2]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PRR4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PRR4
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR