Anti-PTMS Mouse Monoclonal Antibody [clone: 3H5]

Lieferant: Abnova
ABNOH00005763-M17EA 394 EUR
ABNOH00005763-M17
Anti-PTMS Mouse Monoclonal Antibody [clone: 3H5]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant PTMS.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PTMS
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR