Anti-SNRPB Mouse Monoclonal Antibody [clone: 1B4]

Lieferant: Abnova
ABNOH00006628-M01AEA 382 EUR
ABNOH00006628-M01A
Anti-SNRPB Mouse Monoclonal Antibody [clone: 1B4]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant SNRPB.

Type: Primary
Antigen: SNRPB
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR