Anti-OCLN Mouse Monoclonal Antibody [clone: 5A7]

Lieferant: Abnova
ABNOH00004950-M02EA 382 EUR
ABNOH00004950-M02
Anti-OCLN Mouse Monoclonal Antibody [clone: 5A7]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant OCLN.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: OCLN
Clonality: Monoclonal
Clone: 5A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR