Anti-TPT1 Mouse Monoclonal Antibody [clone: 2A3]

Lieferant: Abnova
ABNOH00007178-M06EA 382 EUR
ABNOH00007178-M06
Anti-TPT1 Mouse Monoclonal Antibody [clone: 2A3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TPT1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TPT1
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR