Anti-NFKBIA Mouse Monoclonal Antibody [clone: 3H4]

Lieferant: Abnova
ABNOH00004792-M10EA 382 EUR
ABNOH00004792-M10
Anti-NFKBIA Mouse Monoclonal Antibody [clone: 3H4]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant NFKBIA.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: NFKBIA
Clonality: Monoclonal
Clone: 3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR