Anti-PPP3CC Mouse Monoclonal Antibody [clone: 4D1]

Lieferant: Abnova
ABNOH00005533-M01EA 382 EUR
ABNOH00005533-M01
Anti-PPP3CC Mouse Monoclonal Antibody [clone: 4D1]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PPP3CC.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PPP3CC
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR