Anti-TNFRSF1A Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00007132-D03PEA 368 EUR
ABNOH00007132-D03P
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human TNFRSF1A protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR