Anti-TLN1 Mouse Monoclonal Antibody [clone: 2B11]

Lieferant: Abnova
ABNOH00007094-M80AEA 382 EUR
ABNOH00007094-M80A
Anti-TLN1 Mouse Monoclonal Antibody [clone: 2B11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TLN1.

Type: Primary
Antigen: TLN1
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR