Anti-TLN1 Mouse Monoclonal Antibody [clone: 5D1]

Lieferant: Abnova
ABNOH00007094-M02EA 382 EUR
ABNOH00007094-M02
Anti-TLN1 Mouse Monoclonal Antibody [clone: 5D1]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant TLN1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TLN1
Clonality: Monoclonal
Clone: 5D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR