Anti-RRM1 Mouse Monoclonal Antibody [clone: 2D11]

Lieferant: Abnova
ABNOH00006240-M08EA 382 EUR
ABNOH00006240-M08
Anti-RRM1 Mouse Monoclonal Antibody [clone: 2D11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant RRM1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: RRM1
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR