Anti-RGS10 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00006001-B01PEA 394 EUR
ABNOH00006001-B01P
Anti-RGS10 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human RGS10 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: RGS10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human