Anti-PGR Mouse Monoclonal Antibody [clone: 1B11]

Lieferant: Abnova
ABNOH00005241-M05AEA 382 EUR
ABNOH00005241-M05A
Anti-PGR Mouse Monoclonal Antibody [clone: 1B11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PGR.

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR