Anti-PEX10 Mouse Monoclonal Antibody [clone: 1B8]

Lieferant: Abnova
ABNOH00005192-M01AEA 394 EUR
ABNOH00005192-M01A
Anti-PEX10 Mouse Monoclonal Antibody [clone: 1B8]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant PEX10.

Type: Primary
Antigen: PEX10
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR