Anti-MAP3K5 Mouse Monoclonal Antibody [clone: X1]

Lieferant: Abnova
ABNOH00004217-M06AEA 382 EUR
ABNOH00004217-M06A
Anti-MAP3K5 Mouse Monoclonal Antibody [clone: X1]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant MAP3K5.

Type: Primary
Antigen: MAP3K5
Clonality: Monoclonal
Clone: X1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR