Anti-STAT4 Mouse Monoclonal Antibody [clone: S1]

Lieferant: Abnova
ABNOH00006775-M02AEA 382 EUR
ABNOH00006775-M02A
Anti-STAT4 Mouse Monoclonal Antibody [clone: S1]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant STAT4.

Type: Primary
Antigen: STAT4
Clonality: Monoclonal
Clone: S1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR