Anti-FTL Mouse Monoclonal Antibody [clone: 1C14]

Lieferant: Abnova
ABNOH00002512-M15EA 382 EUR
ABNOH00002512-M15
Anti-FTL Mouse Monoclonal Antibody [clone: 1C14]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant FTL.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FTL
Clonality: Monoclonal
Clone: 1C14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR