Anti-FOS Mouse Monoclonal Antibody [clone: 1H8]

Lieferant: Abnova
ABNOH00002353-M61EA 382 EUR
ABNOH00002353-M61
Anti-FOS Mouse Monoclonal Antibody [clone: 1H8]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant FOS.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FOS
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR