Anti-FOXD1 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00002297-B01PEA 382 EUR
ABNOH00002297-B01P
Anti-FOXD1 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human FOXD1 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: FOXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR