Anti-FECH Mouse Monoclonal Antibody [clone: 3H3]

Lieferant: Abnova
ABNOH00002235-M07EA 382 EUR
ABNOH00002235-M07
Anti-FECH Mouse Monoclonal Antibody [clone: 3H3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant FECH.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FECH
Clonality: Monoclonal
Clone: 3H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR