Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 2D7]

Lieferant: Abnova
ABNOH00001544-M07EA 382 EUR
ABNOH00001544-M07
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 2D7]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CYP1A2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CYP1A2
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR