Anti-CYB5A Mouse Monoclonal Antibody [clone: 1A8]

Lieferant: Abnova
ABNOH00001528-M06AEA 382 EUR
ABNOH00001528-M06A
Anti-CYB5A Mouse Monoclonal Antibody [clone: 1A8]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant CYB5A.

Type: Primary
Antigen: CYB5A
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR