Anti-FANCF Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00002188-B01PEA 394 EUR
ABNOH00002188-B01P
Anti-FANCF Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human FANCF protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: FANCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human