Anti-CTNND2 Mouse Monoclonal Antibody [clone: 1E3]
Lieferant: Abnova
ABNOH00001501-M02EA 394 EUR
ABNOH00001501-M02
Anti-CTNND2 Mouse Monoclonal Antibody [clone: 1E3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CTNND2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CTNND2
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human