Anti-CTBP1 Mouse Monoclonal Antibody [clone: 2G11]

Lieferant: Abnova
ABNOH00001487-M04EA 394 EUR
ABNOH00001487-M04
Anti-CTBP1 Mouse Monoclonal Antibody [clone: 2G11]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant CTBP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CTBP1
Clonality: Monoclonal
Clone: 2G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR