Anti-CD40 Mouse Monoclonal Antibody [clone: 4F11]

Lieferant: Abnova
ABNOH00000958-M26EA 394 EUR
ABNOH00000958-M26
Anti-CD40 Mouse Monoclonal Antibody [clone: 4F11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant CD40.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR