Anti-CD72 Mouse Monoclonal Antibody [clone: 3F3]

Lieferant: Abnova
ABNOMAB0925EA 338 EUR
ABNOMAB0925
Anti-CD72 Mouse Monoclonal Antibody [clone: 3F3]
Antikörper
Mouse monoclonal antibody raised against native CD72.

Type: Primary
Antigen: CD72
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR