Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/4]

Lieferant: Abnova
ABNOMAB0957EA 399 EUR
ABNOMAB0957
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/4]
Antikörper
Mouse monoclonal antibody raised against partial recombinant HLA-G.

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR